Anti-METRN Rat Monoclonal Antibody [clone: 347505]

Supplier: R & D Systems
MAB3475
220020-470EA 577.02 USD
220020-470
Anti-METRN Rat Monoclonal Antibody [clone: 347505]
Antibodies
Type: Primary
Antigen: METRN
Clonality: Monoclonal
Clone: 347505
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR