Anti-RSPO1 Rat Monoclonal Antibody [clone: 852010]

Supplier: R & D Systems
MAB3474
102885-586EA 594.37 USD
102885-586
Anti-RSPO1 Rat Monoclonal Antibody [clone: 852010]
Antibodies
Type: Primary
Antigen: RSPO1
Clonality: Monoclonal
Clone: 852010
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR