Anti-WNT10A Rat Monoclonal Antibody [clone: 342811]

Supplier: R & D Systems
MAB34691
102885-584EA 577.02 USD
102885-584
Anti-WNT10A Rat Monoclonal Antibody [clone: 342811]
Antibodies
Type: Primary
Antigen: WNT10A
Clonality: Monoclonal
Clone: 342811
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR