Anti-PSIP1 Mouse Monoclonal Antibody [clone: 762815]

Supplier: R & D Systems
MAB3468
102885-582EA 577.02 USD
102885-582
Anti-PSIP1 Mouse Monoclonal Antibody [clone: 762815]
Antibodies
Type: Primary
Antigen: PSIP1
Clonality: Monoclonal
Clone: 762815
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR