Anti-SCGB3A2 Rat Monoclonal Antibody [clone: 381707]

Supplier: R & D Systems
MAB3465
220020-467EA 594.37 USD
220020-467
Anti-SCGB3A2 Rat Monoclonal Antibody [clone: 381707]
Antibodies
Type: Primary
Antigen: SCGB3A2
Clonality: Monoclonal
Clone: 381707
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR