Anti-MIP-II Mouse Monoclonal Antibody [clone: 82206]

Supplier: R & D Systems
MAB346
220020-465EA 772.25 USD
220020-465
Anti-MIP-II Mouse Monoclonal Antibody [clone: 82206]
Antibodies
Type: Primary
Antigen: MIP-II
Clonality: Monoclonal
Clone: 82206
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR