Anti-SUMF2 Rat Monoclonal Antibody [clone: 382407]

Supplier: R & D Systems
MAB3454
220020-464EA 561.11 USD
220020-464
Anti-SUMF2 Rat Monoclonal Antibody [clone: 382407]
Antibodies
Type: Primary
Antigen: SUMF2
Clonality: Monoclonal
Clone: 382407
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR