Anti-AMICA1 Mouse Monoclonal Antibody [clone: 4191]

Supplier: R & D Systems
MAB34491
220020-462EA 594.37 USD
220020-462
Anti-AMICA1 Mouse Monoclonal Antibody [clone: 4191]
Antibodies
Type: Primary
Antigen: AMICA1
Clonality: Monoclonal
Clone: 4191
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR