Anti-CHAT Mouse Monoclonal Antibody [clone: 334008]

Supplier: R & D Systems
MAB3447
220020-460EA 577.02 USD
220020-460
Anti-CHAT Mouse Monoclonal Antibody [clone: 334008]
Antibodies
Type: Primary
Antigen: CHAT
Clonality: Monoclonal
Clone: 334008
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR