Anti-MYOC Mouse Monoclonal Antibody [clone: 297817]

Supplier: R & D Systems
MAB3446
220020-459EA 623.29 USD
220020-459
Anti-MYOC Mouse Monoclonal Antibody [clone: 297817]
Antibodies
Type: Primary
Antigen: MYOC
Clonality: Monoclonal
Clone: 297817
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR