Anti-NEUROG3 Mouse Monoclonal Antibody [clone: 258602]

Supplier: R & D Systems
MAB3444
220020-458EA 561.11 USD
220020-458
Anti-NEUROG3 Mouse Monoclonal Antibody [clone: 258602]
Antibodies
Type: Primary
Antigen Symbol: NEUROG3
Antigen Name: neurogenin 3
Antigen Synonyms: Atoh5, Math4B, NGN-3, bHLHa7, ngn3
Clonality: Monoclonal
Clone: 258602
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR