Anti-THOP1 Mouse Monoclonal Antibody [clone: 455710]

Supplier: R & D Systems
MAB3439
220020-456EA 577.02 USD
220020-456
Anti-THOP1 Mouse Monoclonal Antibody [clone: 455710]
Antibodies
Type: Primary
Antigen: THOP1
Clonality: Monoclonal
Clone: 455710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR