Anti-DPP7 Rat Monoclonal Antibody [clone: 398011]

Supplier: R & D Systems
MAB3438
220020-454EA 577.02 USD
220020-454
Anti-DPP7 Rat Monoclonal Antibody [clone: 398011]
Antibodies
Type: Primary
Antigen: DPP7
Clonality: Monoclonal
Clone: 398011
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR