Anti-ACE2 Rat Monoclonal Antibody [clone: 4652]

Supplier: R & D Systems
MAB3437
220020-453EA 594.37 USD
220020-453
Anti-ACE2 Rat Monoclonal Antibody [clone: 4652]
Antibodies
Type: Primary
Antigen: ACE2
Clonality: Monoclonal
Clone: 4652
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR