Anti-KRAS Rat Monoclonal Antibody [clone: 342404]

Supplier: R & D Systems
MAB3429-100
102885-580EA 633.42 USD
102885-580
Anti-KRAS Rat Monoclonal Antibody [clone: 342404]
Antibodies
Type: Primary
Antigen: KRAS
Clonality: Monoclonal
Clone: 342404
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR