Anti-PTPN22 Mouse Monoclonal Antibody [clone: 340113]

Supplier: R & D Systems
MAB3428
220020-450EA 594.37 USD
220020-450
Anti-PTPN22 Mouse Monoclonal Antibody [clone: 340113]
Antibodies
Type: Primary
Antigen: PTPN22
Clonality: Monoclonal
Clone: 340113
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR