Anti-TYK2 Mouse Monoclonal Antibody [clone: 307214]

Supplier: R & D Systems
MAB3427
220020-449EA 577.02 USD
220020-449
Anti-TYK2 Mouse Monoclonal Antibody [clone: 307214]
Antibodies
Type: Primary
Antigen: TYK2
Clonality: Monoclonal
Clone: 307214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR