Anti-RHEB Mouse Monoclonal Antibody [clone: 344912]

Supplier: R & D Systems
MAB3426
220020-448EA 600.15 USD
220020-448
Anti-RHEB Mouse Monoclonal Antibody [clone: 344912]
Antibodies
Type: Primary
Antigen: RHEB
Clonality: Monoclonal
Clone: 344912
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR