Anti-NFKBIB Mouse Monoclonal Antibody [clone: 347428]

Supplier: R & D Systems
MAB3425
220020-447EA 577.02 USD
220020-447
Anti-NFKBIB Mouse Monoclonal Antibody [clone: 347428]
Antibodies
Type: Primary
Antigen Symbol: NFKBIB
Antigen Name: nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta
Antigen Synonyms: IKBB, TRIP9
Clonality: Monoclonal
Clone: 347428
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR