Anti-BRAF Mouse Monoclonal Antibody [clone: 336016]

Supplier: R & D Systems
MAB3424
220020-446EA 566.89 USD
220020-446
Anti-BRAF Mouse Monoclonal Antibody [clone: 336016]
Antibodies
Type: Primary
Antigen: BRAF
Clonality: Monoclonal
Clone: 336016
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR