Anti-SOD3 Mouse Monoclonal Antibody [clone: 713707]

Supplier: R & D Systems
MAB34201
102885-578EA 594.37 USD
102885-578
Anti-SOD3 Mouse Monoclonal Antibody [clone: 713707]
Antibodies
Type: Primary
Antigen: SOD3
Clonality: Monoclonal
Clone: 713707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR