Anti-SOD2 Mouse Monoclonal Antibody [clone: 349810]

Supplier: R & D Systems
MAB3419
220020-444EA 633.42 USD
220020-444
Anti-SOD2 Mouse Monoclonal Antibody [clone: 349810]
Antibodies
Type: Primary
Antigen: SOD2
Clonality: Monoclonal
Clone: 349810
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR