Anti-SOD1 Mouse Monoclonal Antibody [clone: 348808]

Supplier: R & D Systems
MAB3418
220020-443EA 594.37 USD
220020-443
Anti-SOD1 Mouse Monoclonal Antibody [clone: 348808]
Antibodies
Type: Primary
Antigen: SOD1
Clonality: Monoclonal
Clone: 348808
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR