Anti-CD200R1 Mouse Monoclonal Antibody [clone: 380529]

Supplier: R & D Systems
MAB34141
220020-442EA 594.37 USD
220020-442
Anti-CD200R1 Mouse Monoclonal Antibody [clone: 380529]
Antibodies
Type: Primary
Antigen: CD200R1
Clonality: Monoclonal
Clone: 380529
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR