Anti-H2AFX Mouse Monoclonal Antibody [clone: 322105]

Supplier: R & D Systems
MAB3406
220020-440EA 561.11 USD
220020-440
Anti-H2AFX Mouse Monoclonal Antibody [clone: 322105]
Antibodies
Type: Primary
Antigen Symbol: H2AFX
Antigen Name: H2A histone family, member X
Antigen Synonyms: H2A.X, H2A/X, H2AX
Clonality: Monoclonal
Clone: 322105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR