Anti-CIAP Mouse Monoclonal Antibody [clone: 315301]

Supplier: R & D Systems
MAB3400
220020-439EA 639.2 USD
220020-439
Anti-CIAP Mouse Monoclonal Antibody [clone: 315301]
Antibodies
Simple Western

Type: Primary
Antigen: CIAP
Clonality: Monoclonal
Clone: 315301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR