Anti-ACVR2A Mouse Monoclonal Antibody [clone: 12676]

Supplier: R & D Systems
MAB340
220020-438EA 772.25 USD
220020-438
Anti-ACVR2A Mouse Monoclonal Antibody [clone: 12676]
Antibodies
Type: Primary
Antigen: ACVR2A
Clonality: Monoclonal
Clone: 12676
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR