Anti-ACVR2B Mouse Monoclonal Antibody [clone: 642]

Supplier: R & D Systems
MAB3393
220020-437EA 577.02 USD
220020-437
Anti-ACVR2B Mouse Monoclonal Antibody [clone: 642]
Antibodies
Type: Primary
Antigen: ACVR2B
Clonality: Monoclonal
Clone: 642
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR