Anti-MAP2K4 Mouse Monoclonal Antibody [clone: 323042]

Supplier: R & D Systems
MAB3390
220020-434EA 577.02 USD
220020-434
Anti-MAP2K4 Mouse Monoclonal Antibody [clone: 323042]
Antibodies
Type: Primary
Antigen: MAP2K4
Clonality: Monoclonal
Clone: 323042
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR