Anti-SRC Mouse Monoclonal Antibody [clone: 327537]

Supplier: R & D Systems
MAB3389
220020-432EA 577.02 USD
220020-432
Anti-SRC Mouse Monoclonal Antibody [clone: 327537]
Antibodies
Type: Primary
Antigen: SRC
Clonality: Monoclonal
Clone: 327537
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR