Anti-TNFRSF4 Rat Monoclonal Antibody [clone: 443318]

Supplier: R & D Systems
MAB3388
220020-431EA 647.88 USD
220020-431
Anti-TNFRSF4 Rat Monoclonal Antibody [clone: 443318]
Antibodies
Type: Primary
Antigen: TNFRSF4
Clonality: Monoclonal
Clone: 443318
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR