Anti-CD31 Rat Monoclonal Antibody [clone: 377537]

Supplier: R & D Systems
MAB33871
220020-430EA 623.29 USD
220020-430
Anti-CD31 Rat Monoclonal Antibody [clone: 377537]
Antibodies
Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 377537
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR