Anti-CD31 Rat Monoclonal Antibody [clone: 377521]

Supplier: R & D Systems
MAB3387
220020-429EA 577.02 USD
220020-429
Anti-CD31 Rat Monoclonal Antibody [clone: 377521]
Antibodies
Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 377521
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR