Anti-EPHB6 Mouse Monoclonal Antibody [clone: 465327]

Supplier: R & D Systems
MAB33841
220020-428EA 577.02 USD
220020-428
Anti-EPHB6 Mouse Monoclonal Antibody [clone: 465327]
Antibodies
Type: Primary
Antigen: EPHB6
Clonality: Monoclonal
Clone: 465327
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR