Anti-RBP4 Mouse Monoclonal Antibody [clone: 393005]

Supplier: R & D Systems
MAB3378
220020-425EA 577.02 USD
220020-425
Anti-RBP4 Mouse Monoclonal Antibody [clone: 393005]
Antibodies
Type: Primary
Antigen: RBP4
Clonality: Monoclonal
Clone: 393005
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR