Anti-GM12169 Rat Monoclonal Antibody [clone: 339319]

Supplier: R & D Systems
MAB3374
220020-424EA 561.11 USD
220020-424
Anti-GM12169 Rat Monoclonal Antibody [clone: 339319]
Antibodies
Type: Primary
Antigen Symbol: GM12169
Antigen Name: predicted gene 12169
Antigen Synonyms: OTTMUSG00000005497
Clonality: Monoclonal
Clone: 339319
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR