Anti-VEGFB Mouse Monoclonal Antibody [clone: 122627]

Supplier: R & D Systems
MAB3372
220020-423EA 763.57 USD
220020-423
Anti-VEGFB Mouse Monoclonal Antibody [clone: 122627]
Antibodies
Type: Primary
Antigen: VEGFB
Clonality: Monoclonal
Clone: 122627
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR