Anti-VEGFB Mouse Monoclonal Antibody [clone: 67431]

Supplier: R & D Systems
MAB337
220020-421EA 763.57 USD
220020-421
Anti-VEGFB Mouse Monoclonal Antibody [clone: 67431]
Antibodies
Type: Primary
Antigen: VEGFB
Clonality: Monoclonal
Clone: 67431
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR