Anti-CD101 Rat Monoclonal Antibody [clone: 307707]

Supplier: R & D Systems
MAB3368
220020-419EA 647.88 USD
220020-419
Anti-CD101 Rat Monoclonal Antibody [clone: 307707]
Antibodies
Type: Primary
Antigen: CD101
Clonality: Monoclonal
Clone: 307707
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR