Anti-WNT8B Rat Monoclonal Antibody [clone: 341502]

Supplier: R & D Systems
MAB33671
220020-418EA 577.02 USD
220020-418
Anti-WNT8B Rat Monoclonal Antibody [clone: 341502]
Antibodies
Type: Primary
Antigen: WNT8B
Clonality: Monoclonal
Clone: 341502
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR