Anti-WNT8B Rat Monoclonal Antibody [clone: 341504]

Supplier: R & D Systems
MAB3367
220020-417EA 577.02 USD
220020-417
Anti-WNT8B Rat Monoclonal Antibody [clone: 341504]
Antibodies
Type: Primary
Antigen: WNT8B
Clonality: Monoclonal
Clone: 341504
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR