Anti-TAL1 Mouse Monoclonal Antibody [clone: 333803]

Supplier: R & D Systems
MAB3360
220020-413EA 561.11 USD
220020-413
Anti-TAL1 Mouse Monoclonal Antibody [clone: 333803]
Antibodies
Type: Primary
Antigen: TAL1
Clonality: Monoclonal
Clone: 333803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR