Anti-TNC Rat Monoclonal Antibody [clone: 391819]

Supplier: R & D Systems
MAB3358
220020-410EA 588.58 USD
220020-410
Anti-TNC Rat Monoclonal Antibody [clone: 391819]
Antibodies
Type: Primary
Antigen: TNC
Clonality: Monoclonal
Clone: 391819
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR