Anti-DAND5 Rat Monoclonal Antibody [clone: 340531]

Supplier: R & D Systems
MAB3356
220020-409EA 561.11 USD
220020-409
Anti-DAND5 Rat Monoclonal Antibody [clone: 340531]
Antibodies
Type: Primary
Antigen: DAND5
Clonality: Monoclonal
Clone: 340531
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR