Anti-BTLA Rat Monoclonal Antibody [clone: 330104]

Supplier: R & D Systems
MAB3354
220020-408EA 577.02 USD
220020-408
Anti-BTLA Rat Monoclonal Antibody [clone: 330104]
Antibodies
Type: Primary
Antigen: BTLA
Clonality: Monoclonal
Clone: 330104
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR