Anti-PROC Mouse Monoclonal Antibody [clone: 390226]

Supplier: R & D Systems
MAB3349
220020-407EA 577.02 USD
220020-407
Anti-PROC Mouse Monoclonal Antibody [clone: 390226]
Antibodies
Type: Primary
Antigen: PROC
Clonality: Monoclonal
Clone: 390226
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR