Anti-DPCR1 Mouse Monoclonal Antibody [clone: 213201]

Supplier: R & D Systems
MAB3347
220020-405EA 577.02 USD
220020-405
Anti-DPCR1 Mouse Monoclonal Antibody [clone: 213201]
Antibodies
Type: Primary
Antigen: DPCR1
Clonality: Monoclonal
Clone: 213201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR