Anti-CD34 Mouse Monoclonal Antibody [clone: 1H6]

Supplier: R & D Systems
MAB3346
220020-404EA 577.02 USD
220020-404
Anti-CD34 Mouse Monoclonal Antibody [clone: 1H6]
Antibodies
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR