Anti-CCL25 Mouse Monoclonal Antibody [clone: 52529]

Supplier: R & D Systems
MAB3341
220020-403EA 772.25 USD
220020-403
Anti-CCL25 Mouse Monoclonal Antibody [clone: 52529]
Antibodies
Type: Primary
Antigen: CCL25
Clonality: Monoclonal
Clone: 52529
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR