Anti-IL1B Mouse Monoclonal Antibody [clone: 608714]

Supplier: R & D Systems
MAB33401
102885-572EA 561.11 USD
102885-572
Anti-IL1B Mouse Monoclonal Antibody [clone: 608714]
Antibodies
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone: 608714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR